5-Brom-4,6-dichlorpyrimidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

684635-1G
SIAL684635-1GEA 87.3 EUR
SIAL684635-1G
5-Brom-4,6-dichlorpyrimidin, Sigma-Aldrich®
5-Bromo-4,6-dichloropyrimidine
Formel: C₄HBrCl₂N₂
Molecular Weight: 227,88 g/mol
MDL: MFCD00047379
CAS-Nummer: 68797-61-5
EINECS: 272-305-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR