2,2-Difluorpropionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2-Difluoropropanoic acid

M-Clarity™ Qualitätsstufe = MQ100

684686-250MG
SIAL684686-250MGEA 128 EUR
SIAL684686-250MG
2,2-Difluorpropionsäure, Sigma-Aldrich®
2,2-Difluorpropionsäure
Formel: C₃H₄F₂O₂
Molecular Weight: 110,06 g/mol
MDL: MFCD03093764
CAS-Nummer: 373-96-6
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR