4-Brom-2-methylthiobenzamid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

684740-1G
SIAL684740-1GEA 61.9 EUR
SIAL684740-1G
4-Brom-2-methylthiobenzamid, Sigma-Aldrich®
4-Brom-2-methylthiobenzamid
Formel: C₈H₈BrNS
Molecular Weight: 230,12 g/mol
MDL: MFCD09025669
CAS-Nummer: 397845-03-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR