3-Fluor-4-iodpyridin, Sigma-Aldrich®

Lieferant: Merck
Gefahr

685216-5G
SIAL685216-5GEA 674 EUR
SIAL685216-5G
3-Fluor-4-iodpyridin, Sigma-Aldrich®
3-Fluor-4-iodpyridin
Formel: C₅H₃FIN
Molecular Weight: 222.99 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03092927
CAS-Nummer: 22282-75-3
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR