Titan(II)hydrid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Titandihydrid

M-Clarity™ Qualitätsstufe = MQ100

686069-1G
SIAL686069-1GEA 51.7 EUR
SIAL686069-1G
Titan(II)hydrid, Sigma-Aldrich®
Titan(II)hydrid
Formel: H₂Ti
Molecular Weight: 49,9 g/mol
Dichte: 3,91 g/cm³ (20 °C)
MDL: MFCD00151301
CAS-Nummer: 7704-98-5
EINECS: 231-726-8
UN: 1871
ADR: 4.1,II
Merck Index: 14,09473

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR