Scandium(III)chlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
686077-1G
SIAL686077-1GEA
315
EUR
SIAL686077-1G
Scandium(III)chlorid, Sigma-Aldrich®
Scandium(III)chlorid
Formel:
Cl₃Sc Molecular Weight: 151,31 g/mol Schmelzpunkt: 940 °C Dichte: 1,67 g/cm³ (940 °C) |
MDL:
MFCD00011221 CAS-Nummer: 10361-84-9 EINECS: 233-799-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...