2-Amino-5-methyl-1,3,4-thiadiazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

686166-5G
SIAL686166-5GEA 53.1 EUR
SIAL686166-5G
2-Amino-5-methyl-1,3,4-thiadiazol, Sigma-Aldrich®
2-Amino-5-methyl-1,3,4-thiadiazole
Formel: C₃H₅N₃S
Molecular Weight: 115,16 g/mol
MDL: MFCD00003110
CAS-Nummer: 108-33-8
EINECS: 203-573-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR