2,6-Dimethylchinolin-4(1H)-on, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

686174-5G
SIAL686174-5GEA 108 EUR
SIAL686174-5G
2,6-Dimethylchinolin-4(1H)-on, Sigma-Aldrich®
2,6-Dimethylchinolin-4(1H)-on
Formel: C₁₁H₁₁NO
MDL: MFCD00272319
CAS-Nummer: 15644-82-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR