(R)-(+)-4-Chlor-α-methylbenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-4-Chloro-alpha-methylbenzyl Alcohol, (R)-1-(4-Chlorophenyl)ethanol

M-Clarity™ Qualitätsstufe = MQ100

686298-1G
SIAL686298-1GEA 247 EUR
SIAL686298-1G
(R)-(+)-4-Chlor-α-methylbenzylalkohol, Sigma-Aldrich®
(R)-(+)-4-Chlor-α-methylbenzylalkohol
Formel: C₈H₉ClO
Molecular Weight: 156,61 g/mol
MDL: MFCD06797295
CAS-Nummer: 75968-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR