(S)-(-)-4-Brom-α-methylbenzylalkohol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(S)-1-(4-Bromophenyl)ethanol
M-Clarity™ Qualitätsstufe = MQ100
686301-1G
SIAL686301-1GEA
166
EUR
SIAL686301-1G
(S)-(-)-4-Brom-α-methylbenzylalkohol, Sigma-Aldrich®
(S)-(-)-4-Brom-α-methylbenzylalkohol
Formel:
C₈H₉BrO Molecular Weight: 201.06 g/mol |
MDL:
MFCD06201860 CAS-Nummer: 100760-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...