3-Brom-2-hydroxypyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Brom-2-pyridinol

686387-1G
SIAL686387-1GEA 107 EUR
SIAL686387-1G
3-Brom-2-hydroxypyridin, Sigma-Aldrich®
3-Brom-2-hydroxypyridin
Formel: C₅H₄BrNO
Molecular Weight: 174 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03411569
CAS-Nummer: 13466-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR