3-Amino-5-brombenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

686425-5G
SIAL686425-5GEA 192 EUR
SIAL686425-5G
3-Amino-5-brombenzoesäure, Sigma-Aldrich®
3-Amino-5-bromobenzoic acid
Formel: C₇H₆BrNO₂
Molecular Weight: 216,03 g/mol
MDL: MFCD00227745
CAS-Nummer: 42237-85-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR