6-Fluor-o-toluonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

686689-1G
SIAL686689-1GEA 41.6 EUR
SIAL686689-1G
6-Fluor-o-toluonitril, Sigma-Aldrich®
6-Fluor-o-toluonitril
Formel: C₈H₆FN
Molecular Weight: 135,14 g/mol
MDL: MFCD06660277
CAS-Nummer: 198633-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR