2-Fluor-6-methylbenzaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluor-6-tolualdehyd, 6-Fluor-o-tolualdehyd, 6-Fluor-o-tolylaldehyd

M-Clarity™ Qualitätsstufe = MQ100

686719-1G
SIAL686719-1GEA 95 EUR
SIAL686719-1G
2-Fluor-6-methylbenzaldehyd, Sigma-Aldrich®
2-Fluoro-6-methylbenzaldehyde
Formel: C₈H₇FO
Molecular Weight: 138,14 g/mol
MDL: MFCD09258962
CAS-Nummer: 117752-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR