Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 5-amino-1-methyl-1H-pyrazole-4-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

687197-5G
SIAL687197-5GEA 46.7 EUR
SIAL687197-5G
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat, Sigma-Aldrich®
Ethyl-5-amino-1-methyl-1H-pyrazol-4-carboxylat
Formel: C₇H₁₁N₃O₂
Molecular Weight: 169,18 g/mol
MDL: MFCD00051652
CAS-Nummer: 31037-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR