2-(Hydroxymethyl)indol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

687227-1G
SIAL687227-1GEA 70.2 EUR
SIAL687227-1G
2-(Hydroxymethyl)indol, Sigma-Aldrich®
2-(Hydroxymethyl)indole
Formel: C₉H₉NO
MDL: MFCD00030195
CAS-Nummer: 24621-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR