(R)-1-Boc-3-hydroxypiperidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-1-BOC-3-hydroxypiperidine, (R)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, tert-butyl (3R)-3-hydroxypiperidine-1-carboxylate

M-Clarity™ Qualitätsstufe = MQ100

687278-1G
SIAL687278-1GEA 92.9 EUR
SIAL687278-1G
(R)-1-Boc-3-hydroxypiperidin, Sigma-Aldrich®
(R)-1-Boc-3-hydroxypiperidin
Formel: C₁₀H₁₉NO₃
MDL: MFCD04115306
CAS-Nummer: 143900-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR