(S)-1-Boc-3-hydroxypiperidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, (3S)-3-Hydroxypiperidine, N-BOC protected

M-Clarity™ Qualitätsstufe = MQ100

687367-1G
SIAL687367-1GEA 97.4 EUR
SIAL687367-1G
(S)-1-Boc-3-hydroxypiperidin, Sigma-Aldrich®
(S)-1-Boc-3-hydroxypiperidin
Formel: C₁₀H₁₉NO₃
Molecular Weight: 201,27 g/mol
MDL: MFCD04115307
CAS-Nummer: 143900-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR