3-Brom-2-methoxypyridin, Sigma-Aldrich®

Lieferant: Merck

687413-1G
SIAL687413-1GEA 103 EUR
SIAL687413-1G
3-Brom-2-methoxypyridin, Sigma-Aldrich®
3-Bromo-2-methoxypyridine
Formel: C₆H₆BrNO
Molecular Weight: 188,02 g/mol
MDL: MFCD03095349
CAS-Nummer: 13472-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR