(Methylsulfonyl)benzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methylphenylsulfon

68742-10G
SIAL68742-10GEA 169 EUR
SIAL68742-10G
(Methylsulfonyl)benzol, Sigma-Aldrich®
(Methylsulphonyl)benzene
Formel: C₇H₈O₂S
Molecular Weight: 156,21 g/mol
MDL: MFCD00014741
CAS-Nummer: 3112-85-4
EINECS: 221-476-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR