2,4-Difluorresorcin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4-Difluorobenzene-1,3-diol

M-Clarity™ Qualitätsstufe = MQ200

687421-500MG
SIAL687421-500MGEA 181 EUR
SIAL687421-500MG
2,4-Difluorresorcin, Sigma-Aldrich®
2,4-Difluorresorcin
Formel: C₆H₄F₂O₂
CAS-Nummer: 195136-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR