2-Methylisonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methylpyridine-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

687634-1G
SIAL687634-1GEA 80.2 EUR
SIAL687634-1G
2-Methylisonicotinsäure, Sigma-Aldrich®
2-Methylisonicotinsäure
Formel: C₇H₇NO₂
Molecular Weight: 137,14 g/mol
MDL: MFCD00513435
CAS-Nummer: 4021-11-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR