2-Indolcarbaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Indol-2-carboxaldehyd

M-Clarity™ Qualitätsstufe = MQ100

687782-1G
SIAL687782-1GEA 156 EUR
SIAL687782-1G
2-Indolcarbaldehyd, Sigma-Aldrich®
2-Indolcarbaldehyd
Formel: C₉H₇NO
Molecular Weight: 145,16 g/mol
MDL: MFCD03001425
CAS-Nummer: 19005-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR