1-Boc-2-piperidon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Boc-2-Piperidone, 1-N-Boc-2-piperidone

M-Clarity™ Qualitätsstufe = MQ100

688207-5G
SIAL688207-5GEA 80.9 EUR
SIAL688207-5G
1-Boc-2-piperidon, Sigma-Aldrich®
1-Boc-2-piperidon
Formel: C₁₀H₁₇NO₃
Molecular Weight: 199,25 g/mol
MDL: MFCD02179046
CAS-Nummer: 85908-96-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR