Methyl-5-chlorpyrazin-2-carboxylat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 5-chloropyrazine-2-carboxylate

688584-5G
SIAL688584-5GEA 0 EUR
SIAL688584-5G
Methyl-5-chlorpyrazin-2-carboxylat, Sigma-Aldrich®
Methyl-5-chlorpyrazin-2-carboxylat
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172,57 g/mol
MDL: MFCD01632102
CAS-Nummer: 33332-25-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR