Pyrrol-3-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1H-Pyrrole-3-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

689416-250MG
SIAL689416-250MGEA 53.3 EUR
SIAL689416-250MG
Pyrrol-3-carbonsäure, Sigma-Aldrich®
Pyrrol-3-carbonsäure
Formel: C₅H₅NO₂
MDL: MFCD00800594
CAS-Nummer: 931-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR