2-Iod-4-nitrophenol, Sigma-Aldrich®

Lieferant: Merck

689742-1G
SIAL689742-1GEA 389 EUR
SIAL689742-1G
2-Iod-4-nitrophenol, Sigma-Aldrich®
2-Iod-4-nitrophenol
Formel: C₆H₄INO₃
Molecular Weight: 265,01 g/mol
MDL: MFCD01165664
CAS-Nummer: 89487-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR