2-Brom-4-nitrophenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Brom-2-hydroxy-5-nitrobenzol

689858-5G
SIAL689858-5GEA 270 EUR
SIAL689858-5G
2-Brom-4-nitrophenol, Sigma-Aldrich®
2-Brom-4-nitrophenol
Formel: C₆H₄BrNO₃
Molecular Weight: 218.01 g/mol
MDL: MFCD06656567
CAS-Nummer: 5847-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR