o-Xylylendiamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2-Phenylenedimethanamine

690015-1G
SIAL690015-1GEA 423 EUR
SIAL690015-1G
o-Xylylendiamin, Sigma-Aldrich®
o-Xylylendiamin
Formel: C₈H₁₂N₂
Molecular Weight: 136,19 g/mol
MDL: MFCD06212556
CAS-Nummer: 17300-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR