6-Brom-2,2'-bipyridyl, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Bromo-2,2'-bipyridine

M-Clarity™ Qualitätsstufe = MQ100

690066-1G
SIAL690066-1GEA 185 EUR
SIAL690066-1G
6-Brom-2,2'-bipyridyl, Sigma-Aldrich®
6-Brom-2,2'-bipyridyl
Formel: C₁₀H₇BrN₂
Molecular Weight: 235.0800 g/mol
MDL: MFCD01318953
CAS-Nummer: 10495-73-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR