2,7-Dibromnaphthalin, Sigma-Aldrich®

Lieferant: Merck

690171-5G
SIAL690171-5GEA 90.6 EUR
SIAL690171-5G
2,7-Dibromnaphthalin, Sigma-Aldrich®
2,7-Dibromonaphthalene
Formel: C₁₀H₆Br₂
Molecular Weight: 285,97 g/mol
MDL: MFCD00238580
CAS-Nummer: 58556-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR