2-Bromcyclohexanon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

690422-1G
SIAL690422-1GEA 172 EUR
SIAL690422-1G
2-Bromcyclohexanon, Sigma-Aldrich®
2-Bromocyclohexanone
Formel: C₆H₉BrO
CAS-Nummer: 822-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR