1-Decyl-3-methylimidazoliumtetrafluoroborat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Decyl-3-methylimidazolium tetrafluoroborate
M-Clarity™ Qualitätsstufe = MQ100
690708-50G
690708-5G
SIAL690708-50GEA
421
EUR
SIAL690708-50G
SIAL690708-5G
1-Decyl-3-methylimidazoliumtetrafluoroborat, Sigma-Aldrich®
1-Decyl-3-methylimidazoliumtetrafluoroborate
Formel:
C₁₄H₂₇BF₄N₂ Molecular Weight: 310.19 g/mol |
MDL:
MFCD09038853 CAS-Nummer: 244193-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...