1-Decyl-3-methylimidazoliumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Decyl-3-methylimidazolium tetrafluoroborate

M-Clarity™ Qualitätsstufe = MQ100

690708-50G 690708-5G
SIAL690708-50GEA 421 EUR
SIAL690708-50G SIAL690708-5G
1-Decyl-3-methylimidazoliumtetrafluoroborat, Sigma-Aldrich®
1-Decyl-3-methylimidazoliumtetrafluoroborate
Formel: C₁₄H₂₇BF₄N₂
Molecular Weight: 310.19 g/mol
MDL: MFCD09038853
CAS-Nummer: 244193-56-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR