(2-Methoxyethyl)acetoacetat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Methoxyethyl 3-oxobutanoate
M-Clarity™ Qualitätsstufe = MQ100
691062-25KG
SIAL691062-25KGEA
0
EUR
SIAL691062-25KG
(2-Methoxyethyl)acetoacetat, Sigma-Aldrich®
(2-Methoxyethyl)acetoacetat
Formel:
C₇H₁₂O₄ Molecular Weight: 160.1678 g/mol |
MDL:
MFCD00009648 CAS-Nummer: 22502-03-0 EINECS: 245-043-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...