3-Amino-o-anissäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Amino-2-methoxybenzoesäure

M-Clarity™ Qualitätsstufe = MQ100

691429-5G
SIAL691429-5GEA 457 EUR
SIAL691429-5G
3-Amino-o-anissäure, Sigma-Aldrich®
3-Amino-o-anissäure
Formel: C₈H₉NO₃
Molecular Weight: 167,16 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09701221
CAS-Nummer: 861306-04-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR