3-Brom-5-iodpyridin, Sigma-Aldrich®

Lieferant: Merck

691453-1G
SIAL691453-1GEA 65.9 EUR
SIAL691453-1G
3-Brom-5-iodpyridin, Sigma-Aldrich®
3-Bromo-5-iodopyridine
Formel: C₅H₃BrIN
Molecular Weight: 283,89 g/mol
MDL: MFCD03086019
CAS-Nummer: 233770-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR