2-(Trimethylsilyl)benzothiazol, Sigma-Aldrich®

Lieferant: Merck


MQ100

691569-5G
SIAL691569-5GEA 251 EUR
SIAL691569-5G
2-(Trimethylsilyl)benzothiazol, Sigma-Aldrich®
2-(Trimethylsilyl)benzothiazole
Formel: C₆H₁₁NSSi
Molecular Weight: 157.31 g/mol
MDL: MFCD00068185
CAS-Nummer: 32137-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR