(±)-2-Amino-2-(4-chlorphenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: DL-2-(4-Chlorophenyl)glycine, H-DL-Phg(4-Cl)-OH, DL-alpha-Amino-4-chlorophenylacetic acid, 4-Chloro-DL-2-phenylglycine, 4-Chloro-DL-phenylglycine, 2-Amino-2-(4-chlorophenyl)acetic acid

69171-25G-F
SIAL69171-25G-FEA 35.8 EUR
SIAL69171-25G-F
(±)-2-Amino-2-(4-chlorphenyl)essigsäure, Sigma-Aldrich®
(±)-2-Amino-2-(4-chlorphenyl)essigsäure
Formel: C₈H₈ClNO₂
Molecular Weight: 185,61 g/mol
MDL: MFCD00049328
CAS-Nummer: 6212-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR