7-Chloroxindol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 7-Chloroindolin-2-one

M-Clarity™ Qualitätsstufe = MQ100

691712-1G
SIAL691712-1GEA 57.3 EUR
SIAL691712-1G
7-Chloroxindol, Sigma-Aldrich®
7-Chloroxindol
Formel: C₈H₆ClNO
Molecular Weight: 167,5923 g/mol
MDL: MFCD06656001
CAS-Nummer: 25369-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR