4-Chlorisatosäureanhydrid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 7-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione, 7-Chloro-2H-3,1-benzoxazine-2,4(1H)-dione

M-Clarity™ Qualitätsstufe = MQ100

691720-1G
SIAL691720-1GEA 36.4 EUR
SIAL691720-1G
4-Chlorisatosäureanhydrid, Sigma-Aldrich®
4-Chlorisatosäureanhydrid
Formel: C₈H₄ClNO₃
Molecular Weight: 197,58 g/mol
MDL: MFCD00090431
CAS-Nummer: 40928-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR