3,6-Dibrom-2-picolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,6-Dibrom-2-methylpyridin

M-Clarity™ Qualitätsstufe = MQ100

691828-1G
SIAL691828-1GEA 56.6 EUR
SIAL691828-1G
3,6-Dibrom-2-picolin, Sigma-Aldrich®
3,6-Dibrom-2-picolin
Formel: C₆H₅Br₂N
Molecular Weight: 250,92 g/mol
MDL: MFCD06254589
CAS-Nummer: 39919-65-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR