3-Brom-5-hydroxypyridin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: 3-Brom-5-pyridinol

M-Clarity™ Qualitätsstufe = MQ100

691836-1G
SIAL691836-1GEA 61.5 EUR
SIAL691836-1G
3-Brom-5-hydroxypyridin, Sigma-Aldrich®
3-Brom-5-hydroxypyridin
Formel: C₅H₄BrNO
Molecular Weight: 174 g/mol
MDL: MFCD00661305
CAS-Nummer: 74115-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR