2-Amino-5-iodpyrimidin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

691844-5G
SIAL691844-5GEA 210 EUR
SIAL691844-5G
2-Amino-5-iodpyrimidin, Sigma-Aldrich®
2-Amino-5-iodopyrimidine
Formel: C₄H₄IN₃
Molecular Weight: 221 g/mol
MDL: MFCD01075666
CAS-Nummer: 1445-39-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR