2-Nitrophenethylbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Bromoethyl)-2-nitrobenzene

691992-5G
SIAL691992-5GEA 86.7 EUR
SIAL691992-5G
2-Nitrophenethylbromid, Sigma-Aldrich®
2-Nitrophenethylbromid
Formel: C₈H₈BrNO₂
Molecular Weight: 230.06 g/mol
MDL: MFCD06800159
CAS-Nummer: 16793-89-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR