3-(3,5-Difluorphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

692115-5G
SIAL692115-5GEA 160 EUR
SIAL692115-5G
3-(3,5-Difluorphenyl)propionsäure, Sigma-Aldrich®
3-(3,5-Difluorphenyl)propionsäure
Formel: C₉H₈F₂O₂
Molecular Weight: 186,16 g/mol
MDL: MFCD04116054
CAS-Nummer: 84315-24-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR