3-(3,5-Dichlorphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(3,5-Dichlorophenyl)propanoic acid

M-Clarity™ Qualitätsstufe = MQ100

692123-5G
SIAL692123-5GEA 129 EUR
SIAL692123-5G
3-(3,5-Dichlorphenyl)propionsäure, Sigma-Aldrich®
3-(3,5-Dichlorphenyl)propionsäure
Formel: C₉H₈Cl₂O₂
Molecular Weight: 219,07 g/mol
MDL: MFCD04037332
CAS-Nummer: 95333-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR