5-Brom-2-(methylthio)pyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Bromo-2-methylthiopyridine, 2-Methylthio-5-bromopyridine

692891-1G
SIAL692891-1GEA 228 EUR
SIAL692891-1G
5-Brom-2-(methylthio)pyridin, Sigma-Aldrich®
5-Brom-2-(methylthio)pyridin
Formel: C₆H₆BrNS
Molecular Weight: 204,0875 g/mol
MDL: MFCD03411571
CAS-Nummer: 51933-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR