Thulium, Sigma-Aldrich®

Lieferant: Merck

693677-1EA
SIAL693677-1EAEA 1380 EUR
SIAL693677-1EA
Thulium, Sigma-Aldrich®
Thulium
Formel: Tm
Molecular Weight: 168,93 g/mol
MDL: MFCD00011281
CAS-Nummer: 7440-30-4
EINECS: 231-140-2
UN: 3178
ADR: 4.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR