Holmium, Sigma-Aldrich®

Lieferant: Merck

693693-1EA
SIAL693693-1EAEA 687 EUR
SIAL693693-1EA
Holmium, Sigma-Aldrich®
Holmium
Formel: Ho
Molecular Weight: 164,93 g/mol
MDL: MFCD00011049
CAS-Nummer: 7440-60-0
EINECS: 231-169-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR